This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02837863_m1
See other CA6 GE Assays ›
Gene Symbol:
CA6
Gene Name:
carbonic anhydrase 6
Gene Aliases:
-
Chromosome Location:
Chr.1: 219300975 - 219346654 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02837863_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CA6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005544862.2 XP_005544919.1 4 - 5 618 154
XM_015443012.1 XP_015298498.1 4 - 5 607 154
XM_015443016.1 XP_015298502.1 4 - 5 590 154
XM_015443019.1 XP_015298505.1 4 - 5 587 154
XM_015443024.1 XP_015298510.1 4 - 5 612 154
XM_015443028.1 XP_015298514.1 4 - 5 1361 154

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0