This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf02794255_g1
See other G6PD GE Assays ›
Gene Symbol:
G6PD
Gene Name:
glucose-6-phosphate dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.X: 151425276 - 151446998 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf02794255_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: G6PD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005595008.2 XP_005595065.1 3 - 4 640 75
XM_005595010.2 XP_005595067.1 3 - 4 316 75
XM_005595011.2 XP_005595068.1 3 - 4 281 75
XM_005595012.2 XP_005595069.2 3 - 4 357 75
XM_005595013.2 XP_005595070.1 3 - 4 281 75
XM_015444282.1 XP_015299768.1 3 - 4 351 75
XM_015444283.1 XP_015299769.1 3 - 4 359 75
XM_015444284.1 XP_015299770.1 3 - 4 393 75
XM_015444285.1 XP_015299771.1 3 - 4 357 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Inventoried Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0