This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mf00609874_m1
See other FOXN2 GE Assays ›
Gene Symbol:
FOXN2
Gene Name:
forkhead box N2
Gene Aliases:
-
Chromosome Location:
Chr.13: 61899268 - 61964347 on Build M_fas_5.0
Species:
Macaca fascicularis
Species Specific ID (Flybase ID):
-
Assay ID Mf00609874_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FOXN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005575929.2 XP_005575986.1 2 - 3 729 110
XM_005575930.2 XP_005575987.1 3 - 4 864 110
XM_005575931.2 XP_005575988.1 4 - 5 1048 110
XM_005575933.2 XP_005575990.1 2 - 3 661 110
XM_005575934.2 XP_005575991.1 2 - 3 643 110
XM_015433676.1 XP_015289162.1 4 - 5 1092 110
XM_015433678.1 XP_015289164.1 5 - 6 1120 110
XM_015433679.1 XP_015289165.1 4 - 5 1089 110
XM_015433680.1 XP_015289166.1 3 - 4 908 110

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0