This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07291201_m1
See other SRA1 GE Assays ›
Gene Symbol:
SRA1
Gene Name:
steroid receptor RNA activator 1
Gene Aliases:
SRA, SRAP, STRAA1, pp7684
Chromosome Location:
Chr.5: 140550067 - 140558093 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07291201_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SRA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001253764.1 NP_001240693.1 1 - 2 307 57
GenBank mRNA BU553214.1 - 1 - 2 327 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
cell proliferation
cell differentiation
regulation of apoptotic process
positive regulation of receptor activity
transcription coactivator activity
protein binding
ligand-dependent nuclear receptor binding
ligand-dependent nuclear receptor transcription coactivator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0