This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290791_g1
See other FKBP1A GE Assays ›
Gene Symbol:
FKBP1A
Gene Name:
FK506 binding protein 1A
Gene Aliases:
FKBP-12, FKBP-1A, FKBP1, FKBP12, PKC12, PKCI2, PPIASE
Chromosome Location:
Chr.20: 1368977 - 1393172 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290791_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FKBP1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199786.1 NP_001186715.1 2 - 3 259 110
GenBank mRNA AB241124.1 - 2 - 3 85 110
BC119733.1 - 2 - 3 194 110
BU540706.1 - 180 110
CN426120.1 - 2 - 3 163 110

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC119733.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
heart morphogenesis
protein folding
'de novo' protein folding
transforming growth factor beta receptor signaling pathway
SMAD protein complex assembly
protein maturation by protein folding
positive regulation of protein ubiquitination
positive regulation of protein binding
negative regulation of protein phosphatase type 2B activity
regulation of protein localization
regulation of activin receptor signaling pathway
protein refolding
T cell activation
positive regulation of I-kappaB kinase/NF-kappaB signaling
extracellular fibril organization
regulation of immune response
negative regulation of release of sequestered calcium ion into cytosol
ventricular cardiac muscle tissue morphogenesis
regulation of ryanodine-sensitive calcium-release channel activity
negative regulation of ryanodine-sensitive calcium-release channel activity
heart trabecula formation
chaperone-mediated protein folding
calcium ion transmembrane transport
regulation of amyloid precursor protein catabolic process
amyloid fibril formation
peptidyl-prolyl cis-trans isomerase activity
signal transducer activity
transforming growth factor beta receptor binding
protein binding
macrolide binding
FK506 binding
calcium channel inhibitor activity
type I transforming growth factor beta receptor binding
ion channel binding
SMAD binding
activin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0