This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290444_m1
See other ATXN3 GE Assays ›
Gene Symbol:
ATXN3
Gene Name:
ataxin 3
Gene Aliases:
AT3, ATX3, JOS, MJD, MJD1, SCA3
Chromosome Location:
Chr.14: 92058552 - 92106621 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290444_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATXN3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164782.1 NP_001158254.1 1 - 2 94 74
GenBank mRNA GQ176541.1 - 1 - 2 47 74

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001164782.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
nucleotide-excision repair
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
misfolded or incompletely synthesized protein catabolic process
chemical synaptic transmission
nervous system development
regulation of cell-substrate adhesion
protein deubiquitination
actin cytoskeleton organization
cellular response to heat
monoubiquitinated protein deubiquitination
exploration behavior
proteasome-mediated ubiquitin-dependent protein catabolic process
intermediate filament cytoskeleton organization
protein K63-linked deubiquitination
histone H3 deacetylation
protein K48-linked deubiquitination
cellular response to misfolded protein
positive regulation of ERAD pathway
protein localization to cytosolic proteasome complex involved in ERAD pathway
RNA polymerase II regulatory region DNA binding
histone deacetylase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
omega peptidase activity
ubiquitin protein ligase binding
identical protein binding
ATPase binding
Lys63-specific deubiquitinase activity
Lys48-specific deubiquitinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header