This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07290413_m1
See other RPAIN GE Assays ›
Gene Symbol:
RPAIN
Gene Name:
RPA interacting protein
Gene Aliases:
HRIP, RIP
Chromosome Location:
Chr.17: 5419185 - 5433020 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07290413_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPAIN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160266.1 NP_001153738.1 4 - 5 1000 75
GenBank mRNA AA745783.1 - 1 - 2 336 75
N41653.1 - 4 - 5 398 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

DNA-dependent DNA replication
DNA repair
DNA recombination
protein import into nucleus
protein complex binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0