This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07289310_m1
See other UCN3 GE Assays ›
Gene Symbol:
UCN3
Gene Name:
urocortin 3
Gene Aliases:
SCP, SPC, UCNIII
Chromosome Location:
Chr.10: 5365013 - 5374206 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07289310_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCN3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053049.3 NP_444277.2 1 - 2 220 66
GenBank mRNA BC100868.1 - 1 - 2 137 40002995 66
BC100869.1 - 1 - 2 137 40002997 66
BC100870.1 - 1 - 2 137 40002998 66
BM310953.1 - 1 - 2 216 66
BQ776877.1 - 1 - 2 213 66
BQ777111.1 - 1 - 2 223 66

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC100868.1, BC100869.1, BC100870.1, NM_053049.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UCN3

Gene Ontology Categories:

Function(s) Process(es)

response to stress
adenylate cyclase-activating G-protein coupled receptor signaling pathway
digestion
cellular response to nutrient levels
G-protein coupled receptor binding
hormone activity
corticotropin-releasing hormone receptor 2 binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0