This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07288444_m1
See other ARL2 GE Assays ›
Gene Symbol:
ARL2
Gene Name:
ADP ribosylation factor like GTPase 2
Gene Aliases:
ARFL2
Chromosome Location:
Chr.11: 65014113 - 65022185 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07288444_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001667.3 NP_001658.2 3 - 4 435 102
GenBank mRNA AF493888.1 - 3 - 4 340 102
AK223502.1 - 3 - 4 413 102
BC002530.2 - 3 - 4 367 3140402 102
BG706294.1 - 3 - 4 440 102
BT006674.1 - 3 - 4 340 102
L13687.1 - 3 - 4 351 102

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARL2

Gene Ontology Categories:

Function(s) Process(es)

tubulin complex assembly
cell cycle
small GTPase mediated signal transduction
positive regulation of cell-substrate adhesion
regulation of microtubule polymerization
positive regulation of microtubule polymerization
negative regulation of GTPase activity
regulation of insulin secretion
centrosome organization
maintenance of protein location in nucleus
bicellular tight junction assembly
GTPase activity
GTPase inhibitor activity
protein binding
GTP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0