This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287832_m1
See other C1ORF61 GE Assays ›
Gene Symbol:
C1orf61
Gene Name:
chromosome 1 open reading frame 61
Gene Aliases:
CROC4
Chromosome Location:
Chr.1: 156404252 - 156456649 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287832_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C1orf61
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320454.1 NP_001307383.1 1 - 2 210 71
NM_001320455.1 NP_001307384.1 1 - 2 210 71
NR_135267.1 1 - 2 210 71
XM_011509074.1 XP_011507376.1 1 - 2 210 71
GenBank mRNA BF347271.1 - 196 71
HY139418.1 - 1 - 2 210 71
HY152089.1 - 1 - 2 210 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001320454.1, NM_001320455.1, XM_011509074.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0