This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs07287720_m1
See other LYRM4 GE Assays ›
Gene Symbol:
LYRM4
Gene Name:
LYR motif containing 4
Gene Aliases:
C6orf149, CGI-203, COXPD19, ISD11
Chromosome Location:
Chr.6: 5031029 - 5260950 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs07287720_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LYRM4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164840.2 NP_001158312.1 1 - 2 300 74
NM_001164841.2 NP_001158313.1 1 - 2 300 74
NM_001318782.1 NP_001305711.1 1 - 2 300 74
NM_001318783.1 NP_001305712.1 1 - 2 300 74
NM_020408.5 NP_065141.3 1 - 2 300 74
NR_104417.1 1 - 2 300 74
NR_134856.1 1 - 2 300 74
XM_017011083.1 XP_016866572.1 1 - 2 294 74
XM_017011084.1 XP_016866573.1 1 - 2 297 74
GenBank mRNA AF088060.1 - 1 - 2 80 74
AF170070.1 - 1 - 2 96 74
AF285118.1 - 1 - 2 96 74
AK291158.1 - 1 - 2 92 74
AK309661.1 - 1 - 2 102 74
BC009552.2 - 1 - 2 151 3898393 74
BI226933.1 - 1 - 2 274 74
BI753819.1 - 1 - 2 286 74
BI758258.1 - 1 - 2 289 74
BX353584.2 - 1 - 2 79 74
CK819966.1 - 1 - 2 76 74
DA114665.1 - 1 - 2 92 74
DA222333.1 - 1 - 2 92 74
HY168919.1 - 1 - 2 301 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LYRM4

Gene Ontology Categories:

Function(s)

small molecule metabolic process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0