This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06648864_s1
See other MNX1 GE Assays ›
Gene Symbol:
MNX1
Gene Name:
motor neuron and pancreas homeobox 1
Gene Aliases:
HB9, HLXB9, HOXHB9, SCRA1
Chromosome Location:
Chr.7: 157004853 - 157010653 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06648864_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MNX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001165255.1 NP_001158727.1 1 79
GenBank mRNA AY927460.1 - 1 79
AY927462.1 - 3 1033 79
AY927464.1 - 1 79

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001165255.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

helix-turn-helix transcription factor homeodomain transcription factor DNA-binding transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
humoral immune response
anatomical structure morphogenesis
spinal cord motor neuron cell fate specification
endocrine pancreas development
neuron projection morphogenesis
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0