This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06641140_s1
See other FLOT2 GE Assays ›
Gene Symbol:
FLOT2
Gene Name:
flotillin 2
Gene Aliases:
ECS-1, ECS1, ESA, ESA1, M17S1
Chromosome Location:
Chr.17: 28879339 - 28897728 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06641140_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FLOT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004475.2 NP_004466.2 11 1633 69
XM_005257950.3 XP_005258007.1 11 1664 69
XM_005257953.2 XP_005258010.1 10 1568 69
XM_017024394.1 XP_016879883.1 11 1561 69
XM_017024395.1 XP_016879884.1 12 1730 69
XM_017024396.1 XP_016879885.1 11 1567 69
GenBank mRNA AK225491.1 - 4 663 69
BC017292.1 - 10 1455 69
M60922.1 - 11 1489 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FLOT2

Gene Ontology Categories:

Function(s) Process(es)

membrane raft assembly
cell adhesion
chemical synaptic transmission
epidermis development
negative regulation of gene expression
regulation of heterotypic cell-cell adhesion
regulation of toll-like receptor 3 signaling pathway
protein localization to plasma membrane raft
regulation of myoblast differentiation
protein stabilization
establishment of protein localization to plasma membrane
negative regulation of amyloid precursor protein catabolic process
positive regulation of establishment of T cell polarity
protease binding
protein binding
ionotropic glutamate receptor binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0