This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06638638_s1
See other ADGRL1 GE Assays ›
Gene Symbol:
ADGRL1
Gene Name:
adhesion G protein-coupled receptor L1
Gene Aliases:
CIRL1, CL1, LEC2, LPHN1
Chromosome Location:
Chr.19: 14147737 - 14206245 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06638638_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008701.2 NP_001008701.1 6 958 57
NM_014921.4 NP_055736.2 5 943 57
XM_005259818.3 XP_005259875.1 5 999 57
XM_011527796.2 XP_011526098.1 6 1014 57
XM_011527798.2 XP_011526100.1 5 999 57
XM_011527801.2 XP_011526103.1 4 454 57
XM_017026475.1 XP_016881964.1 6 1017 57
XM_017026476.1 XP_016881965.1 7 1127 57
XM_017026477.1 XP_016881966.1 6 1017 57
XM_017026478.1 XP_016881967.1 6 1017 57
XM_017026479.1 XP_016881968.1 6 1017 57
XM_017026480.1 XP_016881969.1 3 432 57
XM_017026481.1 XP_016881970.1 6 1021 57
GenBank mRNA AB020628.1 - 6 960 57
AF307079.1 - 5 927 57
BC028296.1 - 5 702 57
BC052627.1 - 4 684 57

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRL1

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
synaptic vesicle exocytosis
calcium-mediated signaling using intracellular calcium source
positive regulation of synapse assembly
positive regulation of synapse maturation
G-protein coupled receptor activity
protein binding
latrotoxin receptor activity
carbohydrate binding
cell adhesion molecule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0