This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06638040_s1
See other PFAS GE Assays ›
Gene Symbol:
PFAS
Gene Name:
phosphoribosylformylglycinamidine synthase
Gene Aliases:
FGAMS, FGAR-AT, FGARAT, PURL
Chromosome Location:
Chr.17: 8249255 - 8270491 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06638040_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PFAS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012393.2 NP_036525.1 12 1503 59
XM_006721546.2 XP_006721609.1 12 1526 59
XM_017024760.1 XP_016880249.1 11 1305 59
GenBank mRNA AB002359.1 - 12 1462 59
AK292402.1 - 12 1497 59
AK292804.1 - 12 1503 59
AK295895.1 - 10 1421 59
AY545554.1 - 1 28 59
BC146768.1 - 12 1462 9007213 59
BC167158.1 - 12 1462 8862577 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PFAS

Gene Ontology Categories:

Function(s) Process(es)

purine nucleotide biosynthetic process
'de novo' IMP biosynthetic process
glutamine metabolic process
ribonucleoside monophosphate biosynthetic process
purine ribonucleoside monophosphate biosynthetic process
response to drug
phosphoribosylformylglycinamidine synthase activity
ATP binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0