This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06633144_s1
See other ATM GE Assays ›
Gene Symbol:
ATM
Gene Name:
ATM serine/threonine kinase
Gene Aliases:
AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
Chromosome Location:
Chr.11: 108222484 - 108369102 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06633144_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000051.3 NP_000042.3 13 2438 61
XM_005271561.4 XP_005271618.2 14 2499 61
XM_005271562.4 XP_005271619.2 13 2572 61
XM_006718843.3 XP_006718906.1 13 2205 61
XM_011542840.2 XP_011541142.1 14 2293 61
XM_011542842.2 XP_011541144.1 12 2247 61
XM_011542843.2 XP_011541145.1 13 2786 61
XM_011542844.2 XP_011541146.1 13 2388 61
XM_011542845.1 XP_011541147.1 5 901 61
XM_017017789.1 XP_016873278.1 14 2996 61
XM_017017790.1 XP_016873279.1 13 2393 61
XM_017017791.1 XP_016873280.1 13 2786 61
XM_017017792.1 XP_016873281.1 13 2786 61
GenBank mRNA AF035326.1 - 2 232 61
AF035327.1 - 2 208 61
HM437232.1 - 1 158 61
U33841.1 - 14 2242 61
X91196.1 - 13 2786 61

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATM

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA double-strand break processing
DNA synthesis involved in DNA repair
strand displacement
response to hypoxia
somitogenesis
pre-B cell allelic exclusion
DNA replication
DNA repair
double-strand break repair via nonhomologous end joining
protein phosphorylation
cellular response to DNA damage stimulus
DNA damage induced protein phosphorylation
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
telomere maintenance via telomerase
cell cycle arrest
mitotic spindle assembly checkpoint
reciprocal meiotic recombination
signal transduction
brain development
heart development
determination of adult lifespan
intrinsic apoptotic signaling pathway in response to DNA damage
response to ionizing radiation
regulation of autophagy
histone phosphorylation
peptidyl-serine phosphorylation
negative regulation of B cell proliferation
positive regulation of telomere maintenance via telomerase
positive regulation of histone phosphorylation
V(D)J recombination
phosphatidylinositol-3-phosphate biosynthetic process
peptidyl-serine autophosphorylation
lipoprotein catabolic process
regulation of apoptotic process
positive regulation of apoptotic process
positive regulation of DNA damage response, signal transduction by p53 class mediator
positive regulation of neuron apoptotic process
meiotic telomere clustering
protein autophosphorylation
oocyte development
neuron apoptotic process
regulation of telomerase activity
histone mRNA catabolic process
cellular response to gamma radiation
cellular response to X-ray
cellular response to nitrosative stress
signal transduction involved in mitotic G2 DNA damage checkpoint
replicative senescence
establishment of RNA localization to telomere
establishment of macromolecular complex localization to telomere
regulation of cellular response to heat
regulation of signal transduction by p53 class mediator
negative regulation of TORC1 signaling
negative regulation of telomere capping
positive regulation of telomere maintenance via telomere lengthening
positive regulation of telomerase catalytic core complex assembly
DNA binding
protein serine/threonine kinase activity
DNA-dependent protein kinase activity
protein binding
ATP binding
1-phosphatidylinositol-3-kinase activity
protein complex binding
protein dimerization activity
protein N-terminus binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0