This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06626713_s1
See other ARAF GE Assays ›
Gene Symbol:
ARAF
Gene Name:
A-Raf proto-oncogene, serine/threonine kinase
Gene Aliases:
A-RAF, ARAF1, PKS2, RAFA1
Chromosome Location:
Chr.X: 47561100 - 47571921 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06626713_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARAF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256196.1 NP_001243125.1 16 2398 101
NM_001654.4 NP_001645.1 16 2389 101
XM_006724529.2 XP_006724592.1 16 2325 101
XM_011543906.2 XP_011542208.1 16 3715 101
XM_011543908.2 XP_011542210.1 16 3699 101
XM_011543909.2 XP_011542211.1 11 1690 101
GenBank mRNA AB208831.1 - 12 5456 101
AF052136.1 - 6 1096 101
AK130043.1 - 15 1685 101
BC002466.2 - 16 2269 3347509 101
BC007514.2 - 16 2276 2966604 101
X04790.1 - 16 2372 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARAF

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
cellular protein modification process
regulation of TOR signaling
regulation of proteasomal ubiquitin-dependent protein catabolic process
positive regulation of peptidyl-serine phosphorylation
negative regulation of apoptotic process
protein kinase activity
protein serine/threonine kinase activity
receptor signaling protein activity
protein binding
ATP binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0