This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06626318_s1
See other APOB GE Assays ›
Gene Symbol:
APOB
Gene Name:
apolipoprotein B
Gene Aliases:
FLDB, LDLCQ4, apoB-100, apoB-48
Chromosome Location:
Chr.2: 21001429 - 21044073 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06626318_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000384.2 NP_000375.2 26 9405 91
GenBank mRNA AB208846.1 - 1 435 91
HM487065.1 - 26 9277 91
J02610.1 - 26 9355 91
M14162.1 - 26 9277 91
M15421.1 - 3 5374 91
X03045.1 - 1 112 91
X04506.1 - 26 9405 91
X04714.1 - 26 9396 91

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene APOB

Panther Classification:

Molecular Function -

apolipoprotein

Gene Ontology Categories:

Function(s) Process(es)

retinoid metabolic process
in utero embryonic development
triglyceride mobilization
receptor-mediated endocytosis
spermatogenesis
nervous system development
cholesterol metabolic process
fertilization
response to virus
response to carbohydrate
post-embryonic development
response to selenium ion
positive regulation of gene expression
positive regulation of macrophage derived foam cell differentiation
positive regulation of lipid storage
positive regulation of cholesterol storage
triglyceride catabolic process
cholesterol transport
sperm motility
response to lipopolysaccharide
cholesterol efflux
low-density lipoprotein particle remodeling
very-low-density lipoprotein particle assembly
low-density lipoprotein particle clearance
lipoprotein metabolic process
lipoprotein biosynthetic process
lipoprotein catabolic process
cholesterol homeostasis
lipoprotein transport
cellular protein catabolic process
regulation of cholesterol biosynthetic process
artery morphogenesis
leukocyte migration
cellular response to tumor necrosis factor
cellular response to prostaglandin stimulus
protein binding
phospholipid binding
heparin binding
cholesterol transporter activity
lipase binding
low-density lipoprotein particle receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0