This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05649853_s1
See other AES GE Assays ›
Gene Symbol:
AES
Gene Name:
amino-terminal enhancer of split
Gene Aliases:
AES-1, AES-2, ESP1, GRG, GRG5, Grg-5, TLE5
Chromosome Location:
Chr.19: 3052910 - 3063107 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05649853_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AES
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130.5 NP_001121.2 7 141
NM_198969.1 NP_945320.1 7 141
NM_198970.1 NP_945321.1 7 141
XM_006722664.1 XP_006722727.1 7 141
GenBank mRNA AK094591.1 - 7 140
AK095154.1 - 2 141
N23852.1 - 1 135 141

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AES

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
multicellular organism development
organ morphogenesis
negative regulation of gene expression
Wnt signaling pathway
negative regulation of protein binding
negative regulation of transcription, DNA-templated
negative regulation of response to cytokine stimulus
response to interleukin-1
negative regulation of canonical Wnt signaling pathway
positive regulation of anoikis
transcription corepressor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0