This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05644814_s1
See other ARHGDIA GE Assays ›
Gene Symbol:
ARHGDIA
Gene Name:
Rho GDP dissociation inhibitor alpha
Gene Aliases:
GDIA1, HEL-S-47e, NPHS8, RHOGDI, RHOGDI-1
Chromosome Location:
Chr.17: 81867719 - 81871406 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05644814_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGDIA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001185077.2 NP_001172006.1 6 1356 80
NM_001185078.2 NP_001172007.1 6 994 80
NM_001301240.1 NP_001288169.1 7 722 80
NM_001301241.1 NP_001288170.1 7 952 80
NM_001301242.1 NP_001288171.1 7 1014 80
NM_001301243.1 NP_001288172.1 5 1261 80
NM_004309.5 NP_004300.1 6 1126 80
NR_125441.1 5 1049 80
XM_011523574.1 XP_011521876.1 5 1486 80
GenBank mRNA AK293147.1 - 7 992 80
AK300816.1 - 6 967 80
BC005851.2 - 6 1082 2961556 80
BC005875.2 - 6 1082 2961556 80
BC007630.1 - 1 123 80
BC008701.3 - 5 1017 2966760 80
BC009759.2 - 6 1047 3935956 80
BC013846.1 - 6 891 80
BC016031.1 - 5 998 3911525 80
BC016185.1 - 6 1042 4867857 80
BC024258.1 - 6 1042 3027721 80
BC027730.2 - 6 1045 4764249 80
BC028333.1 - 7 967 80
BC075827.1 - 6 1042 6074977 80
BC106044.1 - 6 1085 6162284 80
BF343803.1 - 181 80
BI768191.1 - 7 684 80
BQ674863.1 - 7 636 80
EU794615.1 - 5 1017 80
X69550.1 - 6 1043 80

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARHGDIA

Panther Classification:

Molecular Function -

G-protein modulator

Gene Ontology Categories:

Function(s) Process(es)

movement of cell or subcellular component
negative regulation of cell adhesion
regulation of Rho protein signal transduction
negative regulation of apoptotic process
positive regulation of GTPase activity
negative regulation of axonogenesis
positive regulation of axonogenesis
regulation of small GTPase mediated signal transduction
semaphorin-plexin signaling pathway
regulation of actin cytoskeleton reorganization
Rho GDP-dissociation inhibitor activity
GTPase activator activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header