This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05632079_s1
See other F5 GE Assays ›
Gene Symbol:
F5
Gene Name:
coagulation factor V
Gene Aliases:
FVL, PCCF, RPRGL1, THPH2
Chromosome Location:
Chr.1: 169511954 - 169586632 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05632079_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: F5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000130.4 NP_000121.2 13 4397 166
XM_017000660.1 XP_016856149.1 12 4406 166
GenBank mRNA AJ297255.1 - 1 2277 166
AK226079.1 - 13 4315 166
M14335.1 - 13 4328 166
M16967.1 - 13 4342 166

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene F5

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

platelet degranulation
ER to Golgi vesicle-mediated transport
blood coagulation
blood circulation
platelet activation
COPII vesicle coating
copper ion binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0