This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05630505_s1
See other RGCC GE Assays ›
Gene Symbol:
RGCC
Gene Name:
regulator of cell cycle
Gene Aliases:
C13orf15, RGC-32, RGC32, bA157L14.2
Chromosome Location:
Chr.13: 41457406 - 41470877 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05630505_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RGCC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014059.2 NP_054778.2 2 414 81
GenBank mRNA AF036549.1 - 2 198 81
BC066334.1 - 2 270 5301484 81
DB451897.1 - 2 414 81

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RGCC

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of exit from mitosis
negative regulation of endothelial cell proliferation
positive regulation of extracellular matrix constituent secretion
complement activation
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
negative regulation of cell proliferation
positive regulation of gene expression
positive regulation of epithelial to mesenchymal transition
negative regulation of angiogenesis
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
activation of protein kinase activity
positive regulation of collagen biosynthetic process
negative regulation of blood vessel endothelial cell migration
positive regulation of mitotic nuclear division
positive regulation of transcription from RNA polymerase II promoter
negative regulation of cytokine secretion
positive regulation of cytokine secretion
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of stress fiber assembly
positive regulation of cell cycle arrest
cellular response to hypoxia
mitotic cell cycle arrest
fibroblast activation
negative regulation of fibroblast growth factor production
positive regulation of extracellular matrix assembly
negative regulation of mitotic cell cycle phase transition
negative regulation of cell-cell adhesion mediated by cadherin
positive regulation of endothelial cell apoptotic process
positive regulation of DNA biosynthetic process
protein binding
protein kinase binding
protein kinase activator activity
R-SMAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0