This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05630151_s1
See other SDC3 GE Assays ›
Gene Symbol:
SDC3
Gene Name:
syndecan 3
Gene Aliases:
SDCN, SYND3
Chromosome Location:
Chr.1: 30869466 - 30909735 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05630151_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDC3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014654.3 NP_055469.3 5 3746 92
XM_011542463.1 XP_011540765.1 5 3681 92
XM_011542464.2 XP_011540766.1 5 3951 92
XM_011542466.1 XP_011540768.1 5 3588 92
GenBank mRNA AB007937.1 - 3 5028 92
AL049426.1 - 1 1750 92

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SDC3

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

retinoid metabolic process
glycosaminoglycan biosynthetic process
glycosaminoglycan catabolic process
cell adhesion
cell migration
glycosaminoglycan metabolic process

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0