This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05621165_s1
See other TM9SF3 GE Assays ›
Gene Symbol:
TM9SF3
Gene Name:
transmembrane 9 superfamily member 3
Gene Aliases:
EP70-P-iso, SMBP
Chromosome Location:
Chr.10: 96518110 - 96587052 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05621165_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TM9SF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011539976.2 XP_011538278.1 1 223 78
XM_011539977.1 XP_011538279.1 1 223 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011539976.2, XM_011539977.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0