This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05617657_s1
See other PTPRO GE Assays ›
Gene Symbol:
PTPRO
Gene Name:
protein tyrosine phosphatase, receptor type O
Gene Aliases:
GLEPP1, NPHS6, PTP-OC, PTP-U2, PTPROT, PTPU2, R-PTP-O
Chromosome Location:
Chr.12: 15322257 - 15598331 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05617657_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPRO
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002848.3 NP_002839.1 26 5679 83
NM_030667.2 NP_109592.1 27 5763 83
NM_030668.2 NP_109593.1 14 3270 83
NM_030669.2 NP_109594.1 15 3354 83
NM_030670.2 NP_109595.1 13 4302 83
NM_030671.2 NP_109596.1 14 4386 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTPRO

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
monocyte chemotaxis
regulation of glomerular filtration
negative regulation of glomerular filtration
protein dephosphorylation
axon guidance
negative regulation of cell-substrate adhesion
negative regulation of neuron projection development
lamellipodium assembly
glomerulus development
peptidyl-tyrosine dephosphorylation
slit diaphragm assembly
glomerular visceral epithelial cell differentiation
negative regulation of canonical Wnt signaling pathway
negative regulation of retinal ganglion cell axon guidance
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
protein binding
phosphatase activity
Wnt-protein binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0