This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05612643_s1
See other FGF17 GE Assays ›
Gene Symbol:
FGF17
Gene Name:
fibroblast growth factor 17
Gene Aliases:
FGF-13, FGF-17, HH20
Chromosome Location:
Chr.8: 22039708 - 22048809 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05612643_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGF17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011544683.1 XP_011542985.1 1 121 65
XM_011544685.1 XP_011542987.1 1 121 65

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011544683.1, XM_011544685.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
signal transduction
cell-cell signaling
nervous system development
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
positive regulation of GTPase activity
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
type 1 fibroblast growth factor receptor binding
type 2 fibroblast growth factor receptor binding
growth factor activity
1-phosphatidylinositol-3-kinase activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0