This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05575124_sH
See other VSIG10 GE Assays ›
Gene Symbol:
VSIG10
Gene Name:
V-set and immunoglobulin domain containing 10
Gene Aliases:
-
Chromosome Location:
Chr.12: 118063593 - 118104402 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05575124_sH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VSIG10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019086.5 NP_061959.2 9 2076 89
XM_005253908.4 XP_005253965.1 8 1429 89
XM_006719467.2 XP_006719530.1 8 1773 89
XM_006719468.2 XP_006719531.1 7 1168 89
XM_011538502.1 XP_011536804.1 9 2073 89
XM_017019504.1 XP_016874993.1 10 2090 89
XM_017019505.1 XP_016874994.1 9 1696 89
XM_017019506.1 XP_016874995.1 9 2210 89
GenBank mRNA AK000681.1 - 9 2100 89
BC028698.2 - 9 2020 4838234 89
BC034471.2 - 9 2015 4825667 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VSIG10

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0