This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05486883_s1
See other BACH2 GE Assays ›
Gene Symbol:
BACH2
Gene Name:
BTB domain and CNC homolog 2
Gene Aliases:
BTBD25
Chromosome Location:
Chr.6: 89926528 - 90296912 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05486883_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BACH2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017011156.1 XP_016866645.1 6 1472 94
XM_017011157.1 XP_016866646.1 5 1278 94
XM_017011158.1 XP_016866647.1 7 1551 94
XM_017011159.1 XP_016866648.1 8 1640 94
XM_017011160.1 XP_016866649.1 5 1394 94
XM_017011162.1 XP_016866651.1 4 1301 94
XM_017011163.1 XP_016866652.1 6 1473 94
XM_017011164.1 XP_016866653.1 5 1379 94
XM_017011167.1 XP_016866656.1 6 1472 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017011156.1, XM_017011157.1, XM_017011158.1, XM_017011159.1, XM_017011160.1, XM_017011162.1, XM_017011163.1, XM_017011164.1, XM_017011167.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein ubiquitination
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0