This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05465527_s1
See other MAST4 GE Assays ›
Gene Symbol:
MAST4
Gene Name:
microtubule associated serine/threonine kinase family member 4
Gene Aliases:
-
Chromosome Location:
Chr.5: 66596348 - 67169595 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05465527_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAST4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164664.1 NP_001158136.1 29 6923 77
NM_001290226.1 NP_001277155.1 27 6375 77
NM_001290227.1 NP_001277156.1 26 5946 77
NM_001297651.1 NP_001284580.1 26 6407 77
NM_015183.2 NP_055998.1 28 6116 77
XM_006714606.3 XP_006714669.1 27 6369 77
XM_006714610.2 XP_006714673.1 25 6155 77
XM_011543382.2 XP_011541684.1 31 7015 77
XM_011543384.2 XP_011541686.1 26 8074 77
XM_011543385.2 XP_011541687.1 26 9194 77
XM_011543386.2 XP_011541688.1 26 6025 77
XM_017009447.1 XP_016864936.1 32 7096 77
XM_017009448.1 XP_016864937.1 30 6979 77
XM_017009449.1 XP_016864938.1 27 6414 77
XM_017009450.1 XP_016864939.1 27 6108 77
XM_017009451.1 XP_016864940.1 26 5907 77
XM_017009452.1 XP_016864941.1 27 9423 77
XM_017009453.1 XP_016864942.1 26 16238 77
GenBank mRNA AB002301.1 - 18 5159 77
AY830839.1 - 28 6117 77
BX648796.1 - 3 2954 77
BX648980.1 - 8 3725 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
peptidyl-serine phosphorylation
intracellular signal transduction
magnesium ion binding
protein serine/threonine kinase activity
ATP binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0