This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05433035_s1
See other STK32B GE Assays ›
Gene Symbol:
STK32B
Gene Name:
serine/threonine kinase 32B
Gene Aliases:
HSA250839, STK32, STKG6, YANK2
Chromosome Location:
Chr.4: 5051323 - 5501001 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05433035_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STK32B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017008375.1 XP_016863864.1 7 653 79
XM_017008376.1 XP_016863865.1 7 653 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-serine phosphorylation
intracellular signal transduction
protein serine/threonine kinase activity
ATP binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-55c564d944-rwwh4:80/100.66.128.142:80.
git-commit: 3092084cb0e494f0e07f07633350183de7fa10a8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0