This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05409885_s1
See other PHKA1 GE Assays ›
Gene Symbol:
PHKA1
Gene Name:
phosphorylase kinase regulatory subunit alpha 1
Gene Aliases:
PHKA
Chromosome Location:
Chr.X: 72578814 - 72714181 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05409885_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHKA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122670.1 NP_001116142.1 2 518 122
NM_001172436.1 NP_001165907.1 2 518 122
NM_002637.3 NP_002628.2 2 518 122
XM_006724661.2 XP_006724724.1 2 360 122
GenBank mRNA BC104944.1 - 2 264 8143947 122
BC143499.1 - 2 264 9052007 122
BC143501.1 - 2 264 9052009 122
X73874.1 - 2 378 122

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHKA1

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s) Process(es)

glycogen metabolic process
glycogen catabolic process
generation of precursor metabolites and energy
protein autophosphorylation
phosphorylase kinase activity
calmodulin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0