This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05409618_s1
See other PRKACB GE Assays ›
Gene Symbol:
PRKACB
Gene Name:
protein kinase cAMP-activated catalytic subunit beta
Gene Aliases:
PKA C-beta, PKACB
Chromosome Location:
Chr.1: 84077975 - 84238498 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05409618_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRKACB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001242857.2 NP_001229786.1 13 4079 105
NM_001242858.2 NP_001229787.1 10 4022 105
NM_001242859.2 NP_001229788.1 12 4070 105
NM_001242860.2 NP_001229789.1 13 4214 105
NM_001242861.2 NP_001229790.1 11 4106 105
NM_001242862.2 NP_001229791.1 9 3818 105
NM_002731.3 NP_002722.1 10 4180 105
NM_182948.3 NP_891993.1 10 4084 105
XM_005271015.3 XP_005271072.1 12 4031 105
XM_005271016.1 XP_005271073.1 12 4156 105
XM_005271017.3 XP_005271074.1 11 4021 105
XM_005271018.4 XP_005271075.1 11 4341 105
XM_005271019.1 XP_005271076.1 10 4097 105
XM_006710758.1 XP_006710821.1 12 4170 105
GenBank mRNA AB209189.1 - 13 4150 105
BC035058.1 - 10 4098 5262372 105
BX537705.1 - 10 105
BX641026.1 - 10 4060 105
CR936631.1 - 12 3874 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PRKACB

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
stimulatory C-type lectin receptor signaling pathway
renal water homeostasis
protein phosphorylation
signal transduction
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
blood coagulation
activation of protein kinase A activity
lipoprotein metabolic process
negative regulation of meiotic cell cycle
regulation of protein processing
cellular response to glucagon stimulus
response to clozapine
negative regulation of smoothened signaling pathway involved in dorsal/ventral neural tube patterning
magnesium ion binding
protein serine/threonine kinase activity
cAMP-dependent protein kinase activity
protein binding
ATP binding
kinase activity
ubiquitin protein ligase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header