This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05379853_s1
See other ACVR1 GE Assays ›
Gene Symbol:
ACVR1
Gene Name:
activin A receptor type 1
Gene Aliases:
ACTRI, ACVR1A, ACVRLK2, ALK2, FOP, SKR1, TSRI
Chromosome Location:
Chr.2: 157736446 - 157875880 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05379853_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACVR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011512108.2 XP_011510410.1 1 1178 87

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011512108.2 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

serine/threonine protein kinase receptor

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
patterning of blood vessels
in utero embryonic development
gastrulation with mouth forming second
mesoderm formation
neural crest cell migration
acute inflammatory response
embryonic heart tube morphogenesis
outflow tract septum morphogenesis
atrioventricular valve morphogenesis
mitral valve morphogenesis
endocardial cushion morphogenesis
endocardial cushion fusion
atrial septum primum morphogenesis
protein phosphorylation
transforming growth factor beta receptor signaling pathway
germ cell development
determination of left/right symmetry
negative regulation of signal transduction
positive regulation of pathway-restricted SMAD protein phosphorylation
peptidyl-threonine phosphorylation
signal transduction by protein phosphorylation
regulation of ossification
positive regulation of cell migration
positive regulation of bone mineralization
BMP signaling pathway
activin receptor signaling pathway
negative regulation of activin receptor signaling pathway
positive regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
smooth muscle cell differentiation
pharyngeal system development
pathway-restricted SMAD protein phosphorylation
ventricular septum morphogenesis
cardiac muscle cell fate commitment
BMP signaling pathway involved in heart development
endocardial cushion cell fate commitment
cellular response to glucocorticoid stimulus
cellular response to BMP stimulus
positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
positive regulation of determination of dorsal identity
negative regulation of extrinsic apoptotic signaling pathway
protein kinase activity
protein serine/threonine kinase activity
transmembrane receptor protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
transforming growth factor beta receptor activity, type I
protein binding
ATP binding
activin receptor activity, type I
peptide hormone binding
protein homodimerization activity
SMAD binding
metal ion binding
activin binding
transforming growth factor beta binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0