This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05377060_s1
See other TGFBRAP1 GE Assays ›
Gene Symbol:
TGFBRAP1
Gene Name:
transforming growth factor beta receptor associated protein 1
Gene Aliases:
TRAP-1, TRAP1, VPS3
Chromosome Location:
Chr.2: 105249404 - 105329714 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05377060_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TGFBRAP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011512193.2 XP_011510495.1 12 3306 146

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
intracellular protein transport
signal transduction
transforming growth factor beta receptor signaling pathway
endosome to lysosome transport
endosomal vesicle fusion
transforming growth factor beta receptor binding
protein binding
SMAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0