This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05335367_s1
See other MYO15B GE Assays ›
Gene Symbol:
MYO15B
Gene Name:
myosin XVB
Gene Aliases:
MYO15BP
Chromosome Location:
Chr.17: 75587545 - 75626850 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05335367_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYO15B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001309242.1 NP_001296171.1 1 554 104
XM_017025120.1 XP_016880609.1 1 801 104
XM_017025121.1 XP_016880610.1 1 801 104
XM_017025122.1 XP_016880611.1 1 801 104
XM_017025123.1 XP_016880612.1 1 803 104
XM_017025124.1 XP_016880613.1 1 801 104
XM_017025125.1 XP_016880614.1 1 800 104
XM_017025126.1 XP_016880615.1 1 800 104
XM_017025127.1 XP_016880616.1 1 796 104
XM_017025128.1 XP_016880617.1 1 478 104
XM_017025141.1 XP_016880630.1 1 1067 104
XM_017025142.1 XP_016880631.1 1 1067 104
XM_017025144.1 XP_016880633.1 1 554 104
GenBank mRNA AF418286.1 - 1 554 104
AF418287.1 - 1 554 104
AF418288.1 - 1 554 104
AF418289.1 - 1 554 104
AF418290.1 - 1 554 104
AF418291.1 - 1 554 104

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017025144.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

actin binding motor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0