This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05332853_s1
See other CNP GE Assays ›
Gene Symbol:
CNP
Gene Name:
2',3'-cyclic nucleotide 3' phosphodiesterase
Gene Aliases:
CNP1
Chromosome Location:
Chr.17: 41966741 - 41977736 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05332853_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CNP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033133.4 NP_149124.3 4 4571 72
GenBank mRNA AK098048.1 - 1 1390 72
AK124861.1 - 1 1922 72
BC045561.1 - 1 1922 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CNP

Panther Classification:

Molecular Function -

hydrolase phosphodiesterase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
chemical synaptic transmission
axonogenesis
aging
adult locomotory behavior
cyclic nucleotide catabolic process
response to toxic substance
substantia nigra development
forebrain development
response to lipopolysaccharide
regulation of mitochondrial membrane permeability
oligodendrocyte differentiation
RNA binding
2',3'-cyclic-nucleotide 3'-phosphodiesterase activity
cyclic nucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0