This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05321630_s1
See other GIT1 GE Assays ›
Gene Symbol:
GIT1
Gene Name:
GIT ArfGAP 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 29573469 - 29589592 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05321630_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GIT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001085454.1 NP_001078923.1 21 69
NM_014030.3 NP_054749.2 20 69
XM_011524684.1 XP_011522986.1 21 69
XM_011524685.1 XP_011522987.1 20 69
GenBank mRNA AF124490.1 - 20 69
AK000863.1 - 4 69
AK024826.1 - 15 69
BC005031.1 - 1 69
BC006227.2 - 4 69
BC018998.2 - 3 69
BC032098.1 - 3 69
BC048196.1 - 5 69
BM984210.1 - 1 546 69
BX537553.1 - 12 69
BX537554.1 - 12 69
BX648097.1 - 12 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GIT1

Gene Ontology Categories:

Function(s) Process(es)

regulation of G-protein coupled receptor protein signaling pathway
regulation of cytokinesis
positive regulation of GTPase activity
ephrin receptor signaling pathway
GTPase activator activity
protein binding
protein complex binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0