This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05319420_g1
See other ACTR3BP5 GE Assays ›
Gene Symbol:
ACTR3BP5 ACTR3BP2 ACTR3BP6
Gene Name:
ACTR3B pseudogene 5
ACTR3B pseudogene 2
ACTR3B pseudogene 6
Gene Aliases:
FKSG74
FKSG73
-
Chromosome Location:
Chr.10: 38696596 - 38698241 on Build GRCh38
Chromosome Location:
Chr.2: 91941133 - 91942470 on Build GRCh38
Chromosome Location:
Chr.22: 16486693 - 16488336 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
Assay ID Hs05319420_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACTR3BP5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045000.1 1 1420 169
GenBank mRNA AY026352.1 - 1 1110 169
Gene Symbol: ACTR3BP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027714.1 1 1126 170
GenBank mRNA AY026351.1 - 1 1126 170
Gene Symbol: ACTR3BP6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AY026350.1 - 1 1108 170

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0