This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05312409_g1
See other PDPK1 GE Assays ›
Gene Symbol:
PDPK1
Gene Name:
3-phosphoinositide dependent protein kinase 1
Gene Aliases:
PDK1, PDPK2, PDPK2P, PRO0461
Chromosome Location:
Chr.16: 2537964 - 2603190 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05312409_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDPK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011522521.2 XP_011520823.1 1 3553 102
XM_011522523.2 XP_011520825.1 1 3558 102
XM_017023271.1 XP_016878760.1 1 3558 102
XM_017023275.1 XP_016878764.1 1 3564 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011522521.2, XM_011522523.2, XM_017023271.1, XM_017023275.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

stimulatory C-type lectin receptor signaling pathway
type B pancreatic cell development
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
negative regulation of protein kinase activity
hyperosmotic response
epidermal growth factor receptor signaling pathway
positive regulation of phospholipase activity
regulation of endothelial cell migration
negative regulation of cardiac muscle cell apoptotic process
cell migration
peptidyl-serine phosphorylation
peptidyl-threonine phosphorylation
calcium-mediated signaling
actin cytoskeleton organization
platelet activation
negative regulation of transforming growth factor beta receptor signaling pathway
T cell costimulation
activation of protein kinase B activity
cellular response to insulin stimulus
negative regulation of toll-like receptor signaling pathway
intracellular signal transduction
Fc-epsilon receptor signaling pathway
regulation of I-kappaB kinase/NF-kappaB signaling
regulation of mast cell degranulation
protein autophosphorylation
phosphatidylinositol-mediated signaling
focal adhesion assembly
T cell receptor signaling pathway
positive regulation of release of sequestered calcium ion into cytosol
cellular response to epidermal growth factor stimulus
positive regulation of establishment of protein localization to plasma membrane
extrinsic apoptotic signaling pathway
protein serine/threonine kinase activity
3-phosphoinositide-dependent protein kinase activity
insulin receptor binding
protein binding
ATP binding
phospholipase activator activity
kinase activity
protein kinase binding
phospholipase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0