This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05293814_s1
See other RIN3 GE Assays ›
Gene Symbol:
RIN3
Gene Name:
Ras and Rab interactor 3
Gene Aliases:
-
Chromosome Location:
Chr.14: 92513773 - 92688994 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05293814_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RIN3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001319987.1 NP_001306916.1 9 3278 82
NM_024832.4 NP_079108.3 10 3531 82
XM_011537164.2 XP_011535466.1 9 3266 82
XM_011537165.2 XP_011535467.1 9 3466 82
XM_017021651.1 XP_016877140.1 10 3413 82
XM_017021652.1 XP_016877141.1 9 3407 82
GenBank mRNA AK026092.1 - 5 1977 82
AK128338.1 - 9 3275 82
BC025248.1 - 5 2397 82
BC070062.1 - 11 3288 30332851 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RIN3

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
signal transduction
positive regulation of GTPase activity
GTPase activator activity
protein binding
Rab guanyl-nucleotide exchange factor activity
Rab GTPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0