This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05061419_ft
See other BCR-JAK2 FUSION GE Assays ›
Gene Symbol:
BCR-JAK2 fusion
Gene Name:
FUSION
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05061419_ft
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCR-JAK2 fusion
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA OM_BCR-JAK2_B1J17_COSF987.0 - 71 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Fusion Homo sapiens

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0