Assay Details
Gene Symbol: RGS19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005260182.3 | XP_005260239.1 | 1 | 251 | 53 | |
XM_011528485.2 | XP_011526787.1 | 1 | 241 | 53 | ||
GenBank mRNA | AF285097.1 | - | 1 | 221 | 53 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005260182.3, XM_011528485.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
autophagy
G-protein coupled receptor signaling pathway
small GTPase mediated signal transduction
cell death
dorsal/ventral axis specification
cell differentiation
regulation of catenin import into nucleus
positive regulation of GTPase activity
cellular response to organic cyclic compound
negative regulation of canonical Wnt signaling pathway
Wnt signaling pathway involved in somitogenesis
G-protein alpha-subunit binding
GTPase activator activity
protein binding
beta-catenin binding
protein kinase binding
armadillo repeat domain binding
regulation of transcription, DNA-templated
autophagy
G-protein coupled receptor signaling pathway
small GTPase mediated signal transduction
cell death
dorsal/ventral axis specification
cell differentiation
regulation of catenin import into nucleus
positive regulation of GTPase activity
cellular response to organic cyclic compound
negative regulation of canonical Wnt signaling pathway
Wnt signaling pathway involved in somitogenesis
G-protein alpha-subunit binding
GTPase activator activity
protein binding
beta-catenin binding
protein kinase binding
armadillo repeat domain binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000479996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000479996.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000332298.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000395042.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000479996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000479996.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | ENST00000493165.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_001039467.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_005873.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009852.hg.1 | NM_005873.2 | Refseq |
