This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05052406_g1
See other TCAM1P GE Assays ›
Gene Symbol:
TCAM1P LOC107987248
Gene Name:
testicular cell adhesion molecule 1, pseudogene
intercellular adhesion molecule 1-like
Gene Aliases:
TCAM1
-
Chromosome Location:
Chr.17: 63857016 - 63864379 on Build GRCh38
Chromosome Location:
Chr.17: 63885851 - 63886703 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs05052406_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TCAM1P
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_002947.2 6 1961 109
GenBank mRNA AB026156.1 - 6 1961 109
Gene Symbol: LOC107987248
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017025501.1 XP_016880990.1 2 501 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000463377.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000463377.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000463377.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000466125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000466125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000466125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000466125.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000469014.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000469014.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000469014.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000469014.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000469014.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000476374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000476374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000476374.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000463377.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000463377.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000463377.6 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000466125.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000466125.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000466125.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000466125.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000469014.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000469014.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000469014.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000469014.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000469014.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000476374.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000476374.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000476374.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000478379.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000482005.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 ENST00000485295.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq
Affymetrix Human Clariom D Assay TC1700008578.hg.1 NR_002947.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0