This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05043316_g1
See other LINC01106 GE Assays ›
Gene Symbol:
LINC01106
Gene Name:
long intergenic non-protein coding RNA 1106
Gene Aliases:
-
Chromosome Location:
Chr.2: 110375109 - 110384536 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05043316_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01106
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027244.1 2 1313 85
GenBank mRNA AK095678.1 - 2 1623 85
CR749351.1 - 3 1406 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500008645.hg.1 ENST00000568414.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008645.hg.1 ENST00000568414.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000448359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000448359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000448359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000448359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 ENST00000448359.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000436665.2 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000448359.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000448359.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000448359.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000448359.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 ENST00000448359.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Clariom D Assay TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Clariom D Assay TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Clariom D Assay TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Clariom D Assay TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Clariom D Assay TC0200013869.hg.1 NR_027244.1 Refseq
Affymetrix Human Clariom D Assay TC1500008645.hg.1 ENST00000568414.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008645.hg.1 ENST00000568414.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000336905.3 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 ENST00000419296.1 Ensembl
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046110.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046111.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046112.1 Refseq
Affymetrix Human Clariom D Assay TC0200008830.hg.1 NR_046112.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0