This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05042026_s1
See other CHUK GE Assays ›
Gene Symbol:
CHUK
Gene Name:
conserved helix-loop-helix ubiquitous kinase
Gene Aliases:
IKBKA, IKK-alpha, IKK1, IKKA, NFKBIKA, TCF16
Chromosome Location:
Chr.10: 100188298 - 100229610 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05042026_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHUK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017015611.1 XP_016871100.1 20 2559 70
XM_017015613.1 XP_016871102.1 10 1374 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

stimulatory C-type lectin receptor signaling pathway
skeletal muscle contraction
protein phosphorylation
inflammatory response
immune response
I-kappaB kinase/NF-kappaB signaling
I-kappaB phosphorylation
Rho protein signal transduction
response to virus
response to toxic substance
anatomical structure morphogenesis
response to acetate
regulation of tumor necrosis factor-mediated signaling pathway
response to lipopolysaccharide
response to hydroperoxide
TRIF-dependent toll-like receptor signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
response to drug
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to amino acid
innate immune response
positive regulation of transcription from RNA polymerase II promoter
T cell receptor signaling pathway
positive regulation of NF-kappaB transcription factor activity
striated muscle cell differentiation
stress-activated MAPK cascade
nucleotide-binding oligomerization domain containing signaling pathway
cellular response to tumor necrosis factor
protein kinase activity
protein binding
ATP binding
IkappaB kinase activity
protein homodimerization activity
protein heterodimerization activity
scaffold protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000008637.hg.1 ENST00000443919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008637.hg.1 ENST00000443919.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008637.hg.1 ENST00000443919.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008637.hg.1 ENST00000443919.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0