This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05037009_u1
See other PIP5K1A GE Assays ›
Gene Symbol:
PIP5K1A
Gene Name:
phosphatidylinositol-4-phosphate 5-kinase type 1 alpha
Gene Aliases:
-
Chromosome Location:
Chr.1: 151198003 - 151249536 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05037009_u1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PIP5K1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006711568.3 XP_006711631.2 1 73 104
XM_011510043.2 XP_011508345.1 1 73 104
GenBank mRNA DQ656041.2 - 1 128 101

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Amplicon spans exons and probe does not span exons

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

glycerophospholipid metabolic process
phosphatidylinositol biosynthetic process
phagocytosis
signal transduction
phospholipid biosynthetic process
fibroblast migration
regulation of phosphatidylinositol 3-kinase signaling
cell migration
keratinocyte differentiation
actin cytoskeleton reorganization
phosphatidylinositol phosphorylation
focal adhesion assembly
cell chemotaxis
protein targeting to plasma membrane
activation of GTPase activity
ruffle assembly
1-phosphatidylinositol-3-phosphate 5-kinase activity
protein binding
ATP binding
1-phosphatidylinositol-4-phosphate 5-kinase activity
kinase binding
1-phosphatidylinositol-5-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol-3,4-bisphosphate 5-kinase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0