This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05036152_s1
See other CEP170B GE Assays ›
Gene Symbol:
CEP170B
Gene Name:
centrosomal protein 170B
Gene Aliases:
CEP170R, FAM68C, KIAA0284
Chromosome Location:
Chr.14: 104865313 - 104896770 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05036152_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEP170B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001112726.2 NP_001106197.1 9 1492 132
NM_015005.2 NP_055820.2 8 1360 132
XM_005267550.3 XP_005267607.1 9 1474 132
XM_011536665.2 XP_011534967.1 10 1627 132
XM_011536666.2 XP_011534968.1 9 1990 132
XM_017021227.1 XP_016876716.1 8 2239 132
XM_017021228.1 XP_016876717.1 8 2239 132
XM_017021229.1 XP_016876718.1 8 2244 132
XM_017021230.1 XP_016876719.1 8 2244 132
GenBank mRNA AB006622.3 - 9 1492 132
AL537476.3 - 1 214 132
BC112928.1 - 8 1391 6141521 132
BC127882.1 - 1 132

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CEP170B

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0