This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05029715_u1
See other UIMC1 GE Assays ›
Gene Symbol:
UIMC1
Gene Name:
ubiquitin interaction motif containing 1
Gene Aliases:
RAP80, X2HRIP110
Chromosome Location:
Chr.5: 176905005 - 177022640 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05029715_u1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UIMC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011534568.2 XP_011532870.1 11 1742 103
GenBank mRNA EF077644.1 - 6 517 103

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Amplicon spans exons and probe does not span exons

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair
double-strand break repair via nonhomologous end joining
transcription, DNA-templated
response to ionizing radiation
G2 DNA damage checkpoint
positive regulation of DNA repair
negative regulation of transcription, DNA-templated
histone H2A K63-linked deubiquitination
protein binding
histone binding
K63-linked polyubiquitin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0