This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05022498_s1
See other TOR1AIP2 GE Assays ›
Gene Symbol:
TOR1AIP2
Gene Name:
torsin 1A interacting protein 2
Gene Aliases:
IFRG15, LULL1, NET9
Chromosome Location:
Chr.1: 179839967 - 179878614 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05022498_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TOR1AIP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022347.3 NP_071742.1 3 946 71
XM_017000477.1 XP_016855966.1 2 1619 71
XM_017000478.1 XP_016855967.1 2 952 71
GenBank mRNA AK289793.1 - 3 921 71
BC032790.1 - 1 220 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK289793.1, BC032790.1, NM_022347.3, XM_017000477.1, XM_017000478.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TOR1AIP2

Gene Ontology Categories:

Function(s) Process(es)

endoplasmic reticulum organization
positive regulation of ATPase activity
protein localization to nuclear envelope
ATPase activator activity
ATPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000474318.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000474318.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000483123.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000483123.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000483123.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000495650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000495650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000495650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000495650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000495650.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000367612.7 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000474318.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000474318.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000482587.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000483123.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000483123.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000483123.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000495650.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000495650.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000495650.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000495650.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000495650.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 ENST00000609928.5 Ensembl
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_001199260.1 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_022347.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq
Affymetrix Human Clariom D Assay TC0100016567.hg.1 NM_145034.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0