This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05019057_s1
See other ARHGAP35 GE Assays ›
Gene Symbol:
ARHGAP35
Gene Name:
Rho GTPase activating protein 35
Gene Aliases:
GRF-1, GRLF1, P190-A, P190A, p190ARhoGAP, p190RhoGAP
Chromosome Location:
Chr.19: 46861002 - 47005077 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05019057_s1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP35
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AL831914.1 - 115 155

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000526119.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000526119.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000526119.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000526119.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000526119.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533597.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533597.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533597.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533597.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000534773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000534773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000534773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 ENST00000534773.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000265838.8 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000299355.10 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000524833.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000526119.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000526119.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000526119.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000526119.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000526119.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000527942.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000528370.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531813.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000531853.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000532792.5 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533597.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533597.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533597.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533597.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000533610.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000534773.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000534773.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000534773.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 ENST00000534773.1 Ensembl
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq
Affymetrix Human Clariom D Assay TC1100008983.hg.1 NM_000019.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0